missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ZBTB40 Polyclonal antibody specifically detects ZBTB40 in Human samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | ZBTB40 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | KIAA0478ZNF923, MGC133098, zinc finger and BTB domain containing 40, zinc finger and BTB domain-containing protein 40 |
| Especie del huésped | Rabbit |
| Inmunógeno | zinc finger and BTB domain containing 40. (Uniprot# Q9NUA8) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?