Learn More
WDR68 Antibody (SR1650), Novus Biologicals™
Código de producto. 18692795 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18692795 100 μg 100 microlitros
18612025 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18692795 Proveedor Novus Biologicals N.º de proveedor NBP322492100ul
Rabbit Monoclonal Antibody
WDR68 Monoclonal antibody specifically detects WDR68 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno WDR68
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry
Clasificación Monoclonal
Clon SR1650
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200
Formulación PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gen DDB1 and CUL4 associated factor 7, DDB1- and CUL4-associated factor 7, HAN11AN11, human anthocyanin, seven-WD-repeat protein of the AN11 family-1, SWAN-1, WD repeat domain 68, WD repeat-containing protein 68, WD repeat-containing protein An11 homolog, WDR68, WD-repeat protein
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human WDR68 (Uniprot #: P61962)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cell Biology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 10238
Especies diana Human
Contenido y almacenamiento Store at -20° C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG