missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
VRK3 Polyclonal antibody specifically detects VRK3 in Human samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | VRK3 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | inactive serine/threonine-protein kinase VRK3, serine/threonine-protein kinase VRK3, Serine/threonine-protein pseudokinase VRK3, vaccinia related kinase 3, Vaccinia-related kinase 3 |
| Especie del huésped | Rabbit |
| Inmunógeno | vaccinia related kinase 3. (Uniprot# Q8IV63) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?