missing translation for 'onlineSavingsMsg'
Learn More

VRK3 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30593354 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30593354 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30593354 Supplier Novus Biologicals Supplier No. NBP345580MFV500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VRK3 Polyclonal antibody specifically detects VRK3 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno VRK3
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado mFluor Violet 500 SE
Formulación 50mM Sodium Borate
Alias de gen inactive serine/threonine-protein kinase VRK3, serine/threonine-protein kinase VRK3, Serine/threonine-protein pseudokinase VRK3, vaccinia related kinase 3, Vaccinia-related kinase 3
Especie del huésped Rabbit
Inmunógeno vaccinia related kinase 3. (Uniprot# Q8IV63)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Protein Kinase
Primario o secundario Primary
ID de gen (Entrez) 51231
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.