missing translation for 'onlineSavingsMsg'
Learn More

VPREB3 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Código de producto. 30590783 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30590783 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30590783 Supplier Novus Biologicals Supplier No. NBP345592MFV500

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPREB3 Polyclonal antibody specifically detects VPREB3 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno VPREB3
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado mFluor Violet 500 SE
Formulación 50mM Sodium Borate
Alias de gen N27C7-2, pre-B lymphocyte 3,8HS20, pre-B lymphocyte gene 3, pre-B lymphocyte protein 3, Protein VPreB3
Especie del huésped Rabbit
Inmunógeno pre-B lymphocyte 3. (Uniprot# Q9UKI3)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Immunology
Primario o secundario Primary
ID de gen (Entrez) 29802
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.