missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | VMAT2 |
| Aplicaciones | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | OTI9E11 |
| Conjugado | Alexa Fluor 750 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 |
| Especie del huésped | Mouse |
| Inmunógeno | Full-length protein expressed in 293T cell transfected with human VMAT2 expression vector. |
| Método de purificación | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?