Descripción
TRH-degrading Ectoenzyme/TRHDE Polyclonal antibody specifically detects TRH-degrading Ectoenzyme/TRHDE in Mouse,Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | TRH-degrading Ectoenzyme/TRHDE |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | HRP |
| Formulación | PBS |
| Alias de gen | EC 3.4.19.6, FLJ22381, PAP-IIthyroliberinase, PGPEP2, Pyroglutamyl-peptidase II, Thyroliberinase, thyrotropin-releasing hormone degrading enzyme, thyrotropin-releasing hormone-degrading ectoenzyme, TRH-degrading ectoenzyme, TRH-DEthyrotropin-releasing hormone degrading ectoenzyme, TRH-specific aminopeptidase |
| Especie del huésped | Goat |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE, Arg64-His1025, Accession # Q8K093 |
| Método de purificación | Antigen Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |