Descripción
PEG3 Monoclonal antibody specifically detects PEG3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Especificaciones
Especificaciones
| Antígeno | PEG3 |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | OTI1E10 |
| Conjugado | APC |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | PBS |
| Alias de gen | DKFZp781A095, KIAA0287PW1, paternally expressed 3, paternally expressed gene 3, Zinc finger and SCAN domain-containing protein 24, ZNF904, ZSCAN24Kruppel-type zinc finger protein |
| Especie del huésped | Mouse |
| Inmunógeno | Human recombinant protein fragment corresponding to amino acids 1-364 of human PEG3 (NP_006201) produced in E.coli. |
| Mostrar más |