Descripción
SH2D1A Monoclonal antibody specifically detects SH2D1A in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | SH2D1A |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | S09-5F6 |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Alias de gen | DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthetic peptide of human SH2D1A (Uniprot # O60880) |
| Mostrar más |