Descripción
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for ELISA
Especificaciones
Especificaciones
| Antígeno | Complement Component C2 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 014 |
| Conjugado | PE |
| Dilución | Sandwich ELISA Capture |
| Formulación | PBS |
| Alias de gen | C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C2 (Met 1-Leu 752; Accession#: NP_000054.2). |
| Mostrar más |