Descripción
OR4C15 Polyclonal specifically detects OR4C15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | OR4C15 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Dilución | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulación | 50mM Sodium Borate |
| Alias de gen | olfactory receptor 4C15, Olfactory receptor OR11-127, Olfactory receptor OR11-134, olfactory receptor, family 4, subfamily C, member 15, OR11-127, OR11-134 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human OR4C15 fragment. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Mostrar más |