Descripción
TRAF3IP2 Monoclonal antibody specifically detects TRAF3IP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | TRAF3IP2 |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | 4A3 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulación | In 1x PBS, pH 7.4 |
| N.º de referencia del gen | AAH02823 |
| Alias de gen | ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2 |
| Especie del huésped | Mouse |
| Mostrar más |