Learn More
Angiopoietin-like Protein 1/ANGPTL1 Antibody (OTI1B2), DyLight 755, Novus Biologicals™
Código de producto. 18112302 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18112302 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18112302 Proveedor Novus Biologicals N.º de proveedor NBP271414IR
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
Angiopoietin-like Protein 1/ANGPTL1 Monoclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Angiopoietin-like Protein 1/ANGPTL1
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon OTI1B2
Conjugado DyLight 755
Dilución Western Blot, Immunohistochemistry
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Símbolos de los genes ANGPTL1
Especie del huésped Mouse
Inmunógeno Recombinant protein fragment corresponding to amino acids 24-491 of human Angiopoietin-like Protein 1/ANGPTL1 (NP_004664) produced in E.coli.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Primario o secundario Primary
ID de gen (Entrez) 9068.0
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG2b