Learn More
Myosin Light Chain 2 Antibody (3B2), Alexa Fluor™ 488, Novus Biologicals™
Código de producto. 18090994 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18090994 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18090994 Proveedor Novus Biologicals N.º de proveedor NBP130249AF488
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Myosin Light Chain 2
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon 3B2
Conjugado Alexa Fluor 488
Dilución Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Símbolos de los genes MYL2
Especie del huésped Mouse
Inmunógeno Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423).
Método de purificación Protein G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 4633
Especificidad de la prueba This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein.
Especies diana Human, Mouse, Xenopus species
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG2b κ