Descripción
FFAR2/GPR43 Polyclonal antibody specifically detects FFAR2/GPR43 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot
Especificaciones
Especificaciones
| Antígeno | FFAR2/GPR43 |
| Aplicaciones | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:50-1:100, Immunoprecipitation 1:150, Immunohistochemistry-Paraffin, Dot Blot 1:10000 |
| Formulación | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo- protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Alias de gen | free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43 |
| Especie del huésped | Rabbit |
| Inmunógeno | Synthetic peptide taken within amino acid region 250-300 on human free fatty acid receptor 2 protein. |
| Método de purificación | Affinity-purified |
| Mostrar más |