Learn More
EMAP-II/AIMP1 Antibody, Biotin, Novus Biologicals™
Product Code. 18001619 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18001619 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001619 Supplier Novus Biologicals Supplier No. NBP298343B
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
This item is not returnable. View return policy
Rabbit Polyclonal Antibody
EMAP-II/AIMP1 Polyclonal specifically detects EMAP-II/AIMP1 in Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Antígeno EMAP-II/AIMP1
Aplicaciones Western Blot, ELISA
Clasificación Polyclonal
Conjugado Biotin
Dilución Western Blot, ELISA
Formulación PBS
Alias de gen aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with purified, recombinant EMAP-II/AIMP1 (Ser145-Lys310).
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Cytokine Research
Primario o secundario Primary
ID de gen (Entrez) 9255
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.