missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
USP34 Polyclonal antibody specifically detects USP34 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | USP34 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | Deubiquitinating enzyme 34, EC 3.1.2.15, EC 3.4.19.12, FLJ43910, KIAA0570MGC104459, KIAA0729ubiquitin carboxyl-terminal hydrolase 34, ubiquitin specific peptidase 34, ubiquitin specific protease 34, ubiquitin thioesterase 34, Ubiquitin thiolesterase 34, Ubiquitin-specific-processing protease 34 |
| Especie del huésped | Rabbit |
| Inmunógeno | ubiquitin specific peptidase 34. (Uniprot# Q70CQ2) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?