Learn More
UNC2541, MedChemExpress
Código de producto. 16463316
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16463316 25 mg 25 miligramos
16461457 50 mg 50 miligramos
16457967 5 mg 5 miligramos
16455248 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16463316 Proveedor Medchem Express N.º de proveedor HY125510
MedChemExpress UNC2541 is a potent and Mer tyrosine kinase (MerTK)-specific inhibitor, binds in the MerTK ATP pocket, with an IC50 of 4.4 nM, more selective over Axl, Tyro3 and Flt3. UNC2541 inhibits phosphorylated MerTK (pMerTK; EC50, 510 nM).
UNC2541 is a potent and Mer tyrosine kinase (MerTK)-specific inhibitor, binds in the MerTK ATP pocket, with an IC50 of 4.4 nM, more selective over Axl, Tyro3 and Flt3. UNC2541 inhibits phosphorylated MerTK (pMerTK
Especificaciones
Nombre del producto químico o material UNC2541
CAS 1612782-86-1
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C24H34FN7O2
Cantidad 25 mg
Información de solubilidad DMSO : 10 mg/mL (21.21 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C(C(NCCCC[C@@H]1N)=N2)=CN=C2NCCCCCCNC1=O)NCC3=CC=C(F)C=C3
Peso molecular (g/mol) 471.57
Formula Weight (peso de la fórmula) 471.57
Porcentaje de pureza 99.45%
Grado Research
Forma física Solid