missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TXNL4B Polyclonal antibody specifically detects TXNL4B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | TXNL4B |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | Dim1-like protein, Dim2, DLPthioredoxin-like protein 4B, FLJ20511, thioredoxin-like 4B |
| Especie del huésped | Rabbit |
| Inmunógeno | thioredoxin-like 4B. (Uniprot# Q9NX01) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?