missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TTLL10 Polyclonal antibody specifically detects TTLL10 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Specifications
| Antígeno | TTLL10 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | APC |
| Formulación | PBS |
| Alias de gen | FLJ42131, member 5, tubulin tyrosine ligase-like family, member 10 |
| Especie del huésped | Rabbit |
| Inmunógeno | tubulin tyrosine ligase-like family, member 10. (Uniprot# Q6ZVT0) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?