missing translation for 'onlineSavingsMsg'
Learn More

TSPAN9 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Código de producto. 18498392 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498392 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18498392 Supplier Novus Biologicals Supplier No. NBP177098AF647
 Artículo restringido
Fecha estimada de envoi: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSPAN9 Polyclonal antibody specifically detects TSPAN9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TSPAN9
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Alexa Fluor 647
Formulación 50mM Sodium Borate
Alias de gen NET-5, NET5tetraspanin-9, new EST tetraspan 5, PP1057, Tetraspan NET-5, tetraspanin 9, transmembrane 4 superfamily member tetraspan NET-5, tspan-9
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 15 amino acid synthetic peptide from near the center of human TSPAN9. The immunogen is located within amino acids 90 - 140 of TSPAN9. Amino Acid Squence: MDKVNENAKKDLKEG
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10867
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.