missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRMT5 Polyclonal antibody specifically detects TRMT5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | TRMT5 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | KIAA1393EC 2.1.1.31, M1G-methyltransferase, TRM5 tRNA methyltransferase 5 homolog (S. cerevisiae), TRM5MGC111453, tRNA (guanine-N(1)-)-methyltransferase, tRNA [GM37] methyltransferase, tRNA methyltransferase 5, tRNA-N1G37 methyltransferase |
| Especie del huésped | Rabbit |
| Inmunógeno | TRM5 tRNA methyltransferase 5 homolog. (Uniprot# Q32P41) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?