missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Tollip Polyclonal antibody specifically detects Tollip in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antígeno | Tollip |
| Aplicaciones | Western Blot, ELISA, Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein |
| Especie del huésped | Rabbit |
| Inmunógeno | Antibody was raised against a 16 amino acid synthetic peptide from near the center of human TOLLIP. The immunogen is located within amino acids 70 - 120 of TOLLIP. Amino Acid Squence: ETPTAHNGAKNPRWNK |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?