missing translation for 'onlineSavingsMsg'
Learn More

TMEM50B Antibody [Biotin], Novus Biologicals™

Product Code. 30594786 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30594786 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30594786 Supplier Novus Biologicals Supplier No. NBP345190B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM50B Polyclonal antibody specifically detects TMEM50B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TMEM50B
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen C21orf4, chromosome 21 open reading frame 4, DKFZp686C2482, FLJ26146, HCV p7-transregulated protein 3, HCV p7-trans-regulated protein 3, HCVP7TP3, transmembrane protein 50B
Especie del huésped Rabbit
Inmunógeno transmembrane protein 50B. (Uniprot# P56557)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 757
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.