missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TMEM41A Polyclonal antibody specifically detects TMEM41A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | TMEM41A |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | mFluor Violet 500 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZp686N06149, DKFZp686N107, MGC15397,2900010K02Rik, transmembrane protein 41A |
| Especie del huésped | Rabbit |
| Inmunógeno | transmembrane protein 41A. (Uniprot# Q96HV5) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?