missing translation for 'onlineSavingsMsg'
Learn More

TMEM35 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30594400 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30594400 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30594400 Supplier Novus Biologicals Supplier No. NBP345195V

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMEM35 Polyclonal antibody specifically detects TMEM35 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TMEM35
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado DyLight 405
Formulación 50mM Sodium Borate
Alias de gen FLJ14084, transmembrane protein 35
Especie del huésped Rabbit
Inmunógeno transmembrane protein 35. (Uniprot# Q53FP2)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 59353
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.