missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TMEM214 Polyclonal antibody specifically detects TMEM214 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | TMEM214 |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FLJ20254, FLJ39682, PP446, transmembrane protein 214 |
| Especie del huésped | Rabbit |
| Inmunógeno | transmembrane protein 214. (Uniprot# Q6NUQ4) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?