missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TMCC1 Polyclonal antibody specifically detects TMCC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | TMCC1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 755 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZp686M0169, FLJ42680, KIAA0779, transmembrane and coiled-coil domain family 1, transmembrane and coiled-coil domains 1, transmembrane and coiled-coil domains protein 1 |
| Especie del huésped | Rabbit |
| Inmunógeno | transmembrane and coiled-coil domain family 1. (Uniprot# O94876) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?