Descripción
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize.
Especificaciones
Especificaciones
| Clasificación | Monoclonal |
| Contenido y almacenamiento | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Formulario | Purified |
| Isotype | IgG1 κ |
For Research Use Only