missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TIS11D Polyclonal antibody specifically detects TIS11D in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | TIS11D |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | HF.10, HF10Zinc finger protein HF.10, Zfp105, zinc finger protein 35, zinc finger protein 35 (clone HF.10) |
| Especie del huésped | Rabbit |
| Inmunógeno | zinc finger protein 36, C3H type-like 2. (Uniprot# P47974) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?