missing translation for 'onlineSavingsMsg'
Learn More

TIGD5 Antibody [Biotin], Novus Biologicals™

Código de producto. 30597627 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30597627 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30597627 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP345241B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIGD5 Polyclonal antibody specifically detects TIGD5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TIGD5
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen tigger transposable element derived 5, tigger transposable element-derived protein 5
Especie del huésped Rabbit
Inmunógeno tigger transposable element derived 5. (Uniprot# Q53EQ6)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84948
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.