missing translation for 'onlineSavingsMsg'
Learn More

TAFA1/FAM19A1 Antibody [Janelia Fluor™ 549], Novus Biologicals™

Código de producto. 30734821 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30734821 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30734821 Proveedor Novus Biologicals N.º de proveedor AF5154I

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

TAFA1/FAM19A1 Polyclonal antibody specifically detects TAFA1/FAM19A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antígeno TAFA1/FAM19A1
Aplicaciones Immunohistochemistry, Western Blot, Neutralization
Clasificación Polyclonal
Conjugado Janelia Fluor 549
Formulación 50mM Sodium Borate
Alias de gen Chemokine-like protein TAFA-1, family with sequence similarity 19 (chemokine (C-C motif)-like), member A1, TAFA1, TAFA-1
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human TAFA1/FAM19A1, Ser26-Thr133, Accession # NP_998774
Método de purificación Antigen Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 407738
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.