missing translation for 'onlineSavingsMsg'
Learn More

SUB1 Antibody [DyLight 550], Novus Biologicals Biologicals™

Código de producto. 30511204 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad
30511204 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30511204 Proveedor Novus Biologicals N.º de proveedor NBP338376R

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

SUB1 Polyclonal antibody specifically detects SUB1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SUB1
Aplicaciones ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Clasificación Polyclonal
Conjugado DyLight 550
Formulación 50mM Sodium Borate
Alias de gen activated RNA polymerase II transcription cofactor 4, activated RNA polymerase II transcriptional coactivator p15, MGC102747, p15, PC4p14P15, Positive cofactor 4, RPO2TC1, SUB1 homolog, SUB1 homolog (S. cerevisiae)
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human SUB1 (NP_006704.3).,, Sequence:, MPKSKELVSSSSSGSDSDSEVDKKLKRKKQVAPEKPVKKQKTGETSRALSSSKQSSSSRDDNMFQIGKMRYVSVRDFKGKVLIDIREYWMDPEGEMKPGRKGISLNPEQWSQLKEQISDIDDAVRKL
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación DNA replication Transcription Translation and Splicing
Primario o secundario Primary
ID de gen (Entrez) 10923
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Tipo de producto Antibody
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.