Learn More
Invitrogen™ STAT3 Monoclonal Antibody (9D8)
Descripción
MA1-13042 detects STAT3 from human, mouse, monkey and rat samples. MA1-13042 has been successfully used in Western blot, ChIP, immunofluorescence, immunohistochemistry (paraffin), and immunoprecipitation procedures. By Western blot, this antibody detects a 88 kDa protein corresponding to STAT3. Positive control: HepG2 cells. The MA1-13042 antigen is a recombinant protein corresponding to residues 655-770 of human STAT3. Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay.
Especificaciones
Especificaciones
| Antígeno | STAT3 |
| Aplicaciones | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 9D8 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide |
| génica | STAT3 |
| N.º de referencia del gen | P40763, P42227, P52631 |
| Alias de gen | 1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.