missing translation for 'onlineSavingsMsg'
Learn More

SRPR alpha Antibody, Novus Biologicals™

Código de producto. 18221853 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
250 μg
Tamaño de la unidad:
250 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18221853 250 μg 250 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18221853 Proveedor Novus Biologicals N.º de proveedor NBP266583

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Sheep Polyclonal Antibody

SRPR alpha Polyclonal specifically detects SRPR alpha in Canine samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SRPRalpha
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1-5 ug/ml
Formulación PBS with 0.08% Sodium Azide
N.º de referencia del gen P06625
Alias de gen Docking protein alpha, DP, DP-alpha, MGC17355, MGC3650, MGC9571, signal recognition particle receptor (docking protein), signal recognition particle receptor ('docking protein'), signal recognition particle receptor subunit alpha, Sralpha, SR-alpha, SRP-alpha
Símbolos de los genes SRPRA
Especie del huésped Sheep
Inmunógeno The antibody to SRPR alpha was raised against a recombinant protein corresponding to amino acids 39-295 which includes part of the amino terminal SRPR beta binding region and the hinge region between it and the carboxyl-terminal GTPase domain.
Método de purificación Ammonium sulfate precipitation
Cantidad 250 μg
Primario o secundario Primary
ID de gen (Entrez) 6734
Especies diana Canine
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.