missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SPZ1 Polyclonal antibody specifically detects SPZ1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antígeno | SPZ1 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FLJ25709, NYD-TSP1, spermatogenic leucine zipper 1, spermatogenic leucine zipper protein 1, spermatogenic zip 1, Testis-specific protein 1, Testis-specific protein NYD-TSP1, TSP1 |
| Especie del huésped | Rabbit |
| Inmunógeno | spermatogenic leucine zipper 1. (Uniprot# Q9BXG8) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?