missing translation for 'onlineSavingsMsg'
Learn More

SPANXC Antibody [Biotin], Novus Biologicals™

Código de producto. 30577570 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30577570 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30577570 Proveedor Novus Biologicals N.º de proveedor NBP345849B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

SPANXC Polyclonal antibody specifically detects SPANXC in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SPANXC
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen B1, C, cancer/testis antigen 11.3, cancer/testis antigen family 11, member 3, cancer/testis-associated protein CTp11, cancer/testis-associated protein of 11 kD, cancer-testis-associated protein CTp11, CT11.1, CT11.2, CT11.3, CT11.4, CTp11, dJ171K16.1, NAP-X, nuclear-associated protein SPAN-Xc, SPANX, SPANX family, member C, SPAN-Xa, SPANXA, SPANX-A, SPAN-Xb, SPANXB, SPANX-B, SPANX-C, SPAN-Xc protein, SPANX-D, sperm protein associated with the nucleus on the X chromosome C, sperm protein associated with the nucleus, X chromosome, family member C
Especie del huésped Rabbit
Inmunógeno SPANX family, member C. (Uniprot# Q9NY87)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer
Primario o secundario Primary
ID de gen (Entrez) 64663
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.