Learn More
SorLA Antibody (SR1433), Novus Biologicals™
Código de producto. 18614365 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
18614365 100 μg 100 microlitros
18683315 50 μg 50 microlitros
2 options
Código de producto. 18614365
Marca: Novus Biologicals NBP322079100ul
Rabbit Monoclonal Antibody
SorLA Monoclonal antibody specifically detects SorLA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SorLA
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon SR1433
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500
Formulación PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gen C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human SorLA (Uniprot #: Q92673)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6653
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20° C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG