missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SNX19 Polyclonal antibody specifically detects SNX19 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Especificaciones
Especificaciones
| Antígeno | SNX19 |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 750 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZp667I205, KIAA0254CHET8, sorting nexin 19, sorting nexin-19 |
| Especie del huésped | Rabbit |
| Inmunógeno | sorting nexin 19. (Uniprot# Q92543) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?