missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SMTNL2 Polyclonal antibody specifically detects SMTNL2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | SMTNL2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | FLJ42461, MGC131847, MGC138382, smoothelin-like 2, smoothelin-like protein 2 |
| Especie del huésped | Rabbit |
| Inmunógeno | smoothelin-like 2. (Uniprot# Q2TAL5) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?