missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SMPDL3B Polyclonal antibody specifically detects SMPDL3B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | SMPDL3B |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 350 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | acid sphingomyelinase-like phosphodiesterase 3b, ASML3BASM-like phosphodiesterase 3b, EC 3.1.4.-, sphingomyelin phosphodiesterase, acid-like 3B |
| Especie del huésped | Rabbit |
| Inmunógeno | sphingomyelin phosphodiesterase, acid-like 3B. (Uniprot# Q92485) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?