Learn More
SLU7 Antibody (262), Novus Biologicals™
Código de producto. 18700294 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. unitSize
18700294 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18700294 Proveedor Novus Biologicals N.º de proveedor NBP243793
Mouse Monoclonal Antibody
Ensure accurate, reproducible results in Western Blot
SLU7 Monoclonal specifically detects SLU7 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Dilución Western Blot 1:1000-1:10000
Símbolos de los genes SLU7