missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SLC25A2 Polyclonal antibody specifically detects SLC25A2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | SLC25A2 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | HRP |
| Formulación | PBS |
| Alias de gen | mitochondrial ornithine transporter 2, ORC2, ORNT2, solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 2 |
| Especie del huésped | Rabbit |
| Inmunógeno | solute carrier family 25(mitochondrial carrier; ornithine transporter) member 2. (Uniprot# Q9BXI2) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?