missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
SLC24A4 Polyclonal antibody specifically detects SLC24A4 in Human,Mouse samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | SLC24A4 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PerCP |
| Formulación | PBS |
| Alias de gen | Na(+)/K(+)/Ca(2+)-exchange protein 4, NCKX4FLJ38852, SHEP6, SLC24A2, sodium/potassium/calcium exchanger 4, solute carrier family 24 (sodium/potassium/calcium exchanger), member 4, Solute carrier family 24 member 4 |
| Especie del huésped | Rabbit |
| Inmunógeno | solute carrier family 24(sodium/potassium/calcium exchanger), member 4. (Uniprot# Q8NFF2) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?