missing translation for 'onlineSavingsMsg'
Learn More

SLC1A5 Antibody, Novus Biologicals™

Código de producto. 18740264 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18740264 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18740264 Proveedor Novus Biologicals N.º de proveedor NBP159732

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

SLC1A5 Polyclonal antibody specifically detects SLC1A5 in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SLC1A5
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1.0 ug/ml, Immunohistochemistry 4-8 ug/ml, Immunohistochemistry-Paraffin 4-8 ug/ml
Formulación PBS, 2% Sucrose with 0.09% Sodium Azide
N.º de referencia del gen Q71UA6
Alias de gen AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5
Símbolos de los genes SLC1A5
Especie del huésped Rabbit
Inmunógeno Synthetic peptides corresponding to SLC1A5(solute carrier family 1 (neutral amino acid transporter), member 5) The peptide sequence was selected from the middle region of SLC1A5 (NP_005619). Peptide sequence FGVALRKLGPEGELLIRFFNSFNEATMVLVSWIMWYAPVGIMFLVAGKIV The peptide sequence for this immunogen was taken from within the described region.
Peso molecular del antígeno 60 kDa
Método de purificación Protein A purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6510
Especificidad de la prueba Zebrafish 86%.
Reconstitución Reconstitute in 100μL distilled water to a final antibody concentration of 1mg/mL.
Especies diana Human, Bovine, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.