missing translation for 'onlineSavingsMsg'
Learn More

SERPINE3 Antibody, R&D Systems™

Código de producto. 18498911 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
25ul
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498911 25ul 25 microlitros
18128471 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18498911 Proveedor Novus Biologicals N.º de proveedor NBP21475125ul
  • ONLINE EXCLUSIVE PRICE
    415.00€
    391.65€ / 25 microlitros
    Ahorro 23.35€
    5% Descuento
 Artículo restringido
Fecha estimada de envoi: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

SERPINE3 Polyclonal specifically detects SERPINE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SERPINE3
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen SERPINE3 serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 3
Símbolos de los genes SERPINE3
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to the amino acids: VLELPYLGSAVSLFLVLPRDKDTPLSHIEPHLTASTIHLWTTSLRRARMDVFLPRFRIQNQFNLKSILNSWGVTDLFDPLKANLKGI
Método de purificación Affinity Purified
Cantidad 25ul
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 647174
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.