Learn More
SEA0400, MedChemExpress
Código de producto. 16479157
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
2 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16479157 5 mg 5 miligramos
16476588 10 mg 10 miligramos
16430246 2 mg 2 miligramos
16402687 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16479157 Proveedor Medchem Express N.º de proveedor HY15515
MedChemExpress SEA0400 is a novel and selective inhibitor of the Na+-Ca2+ exchanger (NCX), inhibiting Na+-dependent Ca2+ uptake in cultured neurons, astrocytes, and microglia with IC50s of from 5 to 33 nM.
SEA0400 is a novel and selective inhibitor of the Na+-Ca2+ exchanger (NCX), inhibiting Na+-dependent Ca2+ uptake in cultured neurons, astrocytes, and microglia with IC50s of from 5 to 33 nM.
Especificaciones
Nombre del producto químico o material SEA0400
CAS 223104-29-8
Color Caqui
Notas de grado de pureza Research
Fórmula molecular C21H19F2NO3
Cantidad 5 mg
Información de solubilidad DMSO : ≥ 32 mg/mL (86.17 mM)
SMILES NC1=CC(OCC)=CC=C1OC2=CC=C(OCC3=CC(F)=CC=C3F)C=C2
Peso molecular (g/mol) 371.38
Formula Weight (peso de la fórmula) 371.38
Porcentaje de pureza 98.82%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation