missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid/Spike Protein (RBD) Recombinant Protein
Descripción
Animal Origin Free Purity is >95% by SDS-PAGE.
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
Especificaciones
Especificaciones
| Concentración | 1.5 mg/mL |
| Para utilizar con (aplicación) | Control, ELISA Standard, Binding Assay |
| Formulación | PBS with no preservative; pH 7.4 |
| Nombre | SARS-CoV-2 Nucleocapsid/Spike Protein (RBD) |
| Intervalo de pH | 7.4 |
| Método de purificación | Purified |
| Cantidad | 100 μg |
| Fuente | E. coli |
| Requisitos de almacenamiento | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Estado normativo | RUO |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?