missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RPS16 Polyclonal antibody specifically detects RPS16 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | RPS16 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | PerCP |
| Formulación | PBS |
| Alias de gen | ribosomal protein S16,40S ribosomal protein S16 |
| Especie del huésped | Rabbit |
| Inmunógeno | ribosomal protein S16. (Uniprot# P62249) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?