missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RNF41 Polyclonal antibody specifically detects RNF41 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | RNF41 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | mFluor Violet 450 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | E3 ubiquitin-protein ligase NRDP1, EC 6.3.2, EC 6.3.2.-, FLRF, MGC45228, NRDP1, ring finger protein 41neuregulin receptor degradation protein-1, SBBI03 |
| Especie del huésped | Rabbit |
| Inmunógeno | ring finger protein 41. (Uniprot# Q9H4P4) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?